siRNA Details
Gene Symbol: DMRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130865.2 | NP_001124337.1 | 6 | 1765 | ||
NM_006557.6 | NP_006548.1 | 4 | 2326 | |||
NM_181872.4 | NP_870987.2 | 4 | 1558 | |||
XM_011517687.1 | XP_011515989.1 | 6 | 4106 | |||
XM_011517690.2 | XP_011515992.1 | 5 | 2186 | |||
XM_011517694.2 | XP_011515996.1 | 3 | 1717 | |||
XM_017014213.1 | XP_016869702.1 | 6 | 2508 | |||
XM_017014214.1 | XP_016869703.1 | 5 | 2183 | |||
XM_017014215.1 | XP_016869704.1 | 4 | 2027 | |||
GenBank mRNA | AF284223.1 | - | 3 | 1853 | ||
AF284224.1 | - | 4 | 1436 | |||
AF284225.1 | - | 3 | 1358 | |||
AK290794.1 | - | 6 | 2071 | |||
BC030204.1 | - | 6 | 1765 | |||
BC136493.1 | - | 3 | 1437 | 9020482 | ||
BX647565.1 | - | 5 | 1372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
