siRNA Details
Gene Symbol: TMEM63B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318792.1 | NP_001305721.1 | 11 | 1019 | ||
NM_018426.2 | NP_060896.1 | 11 | 985 | |||
XM_005249211.3 | XP_005249268.1 | 13 | 1365 | |||
XM_005249213.4 | XP_005249270.1 | 12 | 1292 | |||
XM_005249214.4 | XP_005249271.1 | 10 | 879 | |||
XM_005249215.4 | XP_005249272.1 | 11 | 1184 | |||
XM_005249217.1 | XP_005249274.1 | 11 | 1057 | |||
XM_006715135.1 | XP_006715198.1 | 11 | 1041 | |||
XM_017010999.1 | XP_016866488.1 | 13 | 1242 | |||
XM_017011000.1 | XP_016866489.1 | 12 | 1223 | |||
XM_017011001.1 | XP_016866490.1 | 10 | 923 | |||
XM_017011002.1 | XP_016866491.1 | 10 | 1026 | |||
GenBank mRNA | AK093629.1 | - | 11 | 1041 | ||
AL834238.1 | - | 4 | 283 | |||
BC022095.1 | - | 8 | 622 | |||
BC136769.1 | - | 10 | 861 | 9020759 | ||
CR627460.1 | - | 11 | 1022 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
