siRNA Details
Gene Symbol: TMEM63B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318792.1 | NP_001305721.1 | 10 | 937 | ||
NM_018426.2 | NP_060896.1 | 10 | 903 | |||
XM_005249211.3 | XP_005249268.1 | 12 | 1283 | |||
XM_005249213.4 | XP_005249270.1 | 11 | 1210 | |||
XM_005249214.4 | XP_005249271.1 | 9 | 797 | |||
XM_005249215.4 | XP_005249272.1 | 10 | 1102 | |||
XM_005249217.1 | XP_005249274.1 | 10 | 975 | |||
XM_006715135.1 | XP_006715198.1 | 10 | 959 | |||
XM_017010999.1 | XP_016866488.1 | 12 | 1160 | |||
XM_017011000.1 | XP_016866489.1 | 11 | 1141 | |||
XM_017011001.1 | XP_016866490.1 | 9 | 841 | |||
XM_017011002.1 | XP_016866491.1 | 9 | 944 | |||
GenBank mRNA | AK093629.1 | - | 10 | 959 | ||
AL834238.1 | - | 3 | 201 | |||
BC022095.1 | - | 7 | 540 | |||
BC136769.1 | - | 9 | 779 | 9020759 | ||
CR627460.1 | - | 10 | 940 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
