siRNA Details
Gene Symbol: HIST1H2AB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003513.2 | NP_003504.2 | 1 | 340 | ||
GenBank mRNA | AK311785.1 | - | 1 | 391 | ||
BC125140.1 | - | 1 | 344 | 40119630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
