siRNA Details
Gene Symbol: MDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014641.2 | NP_055456.2 | 8 | 3221 | ||
XM_005249492.2 | XP_005249549.1 | 8 | 3125 | |||
XM_005249493.3 | XP_005249550.1 | 8 | 3122 | |||
XM_005249494.4 | XP_005249551.1 | 8 | 3524 | |||
XM_011515001.2 | XP_011513303.1 | 8 | 2952 | |||
XM_011515003.2 | XP_011513305.1 | 8 | 2807 | |||
XM_011515004.2 | XP_011513306.1 | 9 | 2836 | |||
XM_017011519.1 | XP_016867008.1 | 8 | 2904 | |||
XM_017011520.1 | XP_016867009.1 | 8 | 2820 | |||
GenBank mRNA | BC152556.1 | - | 7 | 2794 | 9007258 | |
D79992.1 | - | 7 | 2794 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
