siRNA Details
Gene Symbol: GTF2H4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001517.4 | NP_001508.1 | 9 | 962 | ||
GenBank mRNA | AK222607.1 | - | 9 | 866 | ||
AK298058.1 | - | 9 | 1010 | |||
AK300239.1 | - | 8 | 815 | |||
AK309869.1 | - | 9 | 976 | |||
BC004935.1 | - | 9 | 934 | 3606718 | ||
BC016302.2 | - | 9 | 934 | 3830902 | ||
BT007321.1 | - | 8 | 755 | |||
Y07595.1 | - | 9 | 882 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
