siRNA Details
Gene Symbol: TRIM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006778.3 | NP_006769.2 | 4, 5 | 854 | ||
NM_052828.2 | NP_439893.2 | 4, 5 | 854 | |||
XM_011514221.1 | XP_011512523.1 | 6, 7 | 1102 | |||
XM_011514222.2 | XP_011512524.1 | 6, 7 | 1336 | |||
XM_011514223.2 | XP_011512525.1 | 7, 8 | 1217 | |||
XM_011514225.1 | XP_011512527.1 | 6, 7 | 1102 | |||
GenBank mRNA | AF220122.1 | - | 4, 5 | 852 | ||
AF220123.1 | - | 4, 5 | 852 | |||
AL360163.1 | - | 2, 3 | 117 | |||
AL360164.1 | - | 2, 3 | 148 | |||
BC093926.1 | - | 4, 5 | 832 | 7939771 | ||
BC113419.1 | - | 4, 5 | 832 | 8322471 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011280, AF134811, AF220121, BC051632, AK011082.
Gene Ontology Categories:
