siRNA Details
Gene Symbol: TRIM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006778.3 | NP_006769.2 | 2, 3 | 596 | ||
NM_052828.2 | NP_439893.2 | 2, 3 | 596 | |||
XM_011514221.1 | XP_011512523.1 | 4, 5 | 844 | |||
XM_011514222.2 | XP_011512524.1 | 4, 5 | 1078 | |||
XM_011514223.2 | XP_011512525.1 | 5, 6 | 959 | |||
XM_011514225.1 | XP_011512527.1 | 4, 5 | 844 | |||
GenBank mRNA | AF220122.1 | - | 2, 3 | 594 | ||
AF220123.1 | - | 2, 3 | 594 | |||
BC093926.1 | - | 2, 3 | 574 | 7939771 | ||
BC113419.1 | - | 2, 3 | 574 | 8322471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
