siRNA Details
Gene Symbol: TRIM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006778.3 | NP_006769.2 | 2 | 571 | ||
NM_052828.2 | NP_439893.2 | 2 | 571 | |||
XM_011514221.1 | XP_011512523.1 | 4 | 819 | |||
XM_011514222.2 | XP_011512524.1 | 4 | 1053 | |||
XM_011514223.2 | XP_011512525.1 | 5 | 934 | |||
XM_011514225.1 | XP_011512527.1 | 4 | 819 | |||
GenBank mRNA | BC093926.1 | - | 2 | 549 | 7939771 | |
BC113419.1 | - | 2 | 549 | 8322471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
