siRNA Details
Gene Symbol: NUSAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243142.1 | NP_001230071.1 | 11 | 2344 | ||
NM_001243143.1 | NP_001230072.1 | 11 | 2305 | |||
NM_001243144.1 | NP_001230073.1 | 9 | 2161 | |||
NM_001301136.1 | NP_001288065.1 | 11 | 2347 | |||
NM_016359.4 | NP_057443.2 | 11 | 2350 | |||
NM_018454.7 | NP_060924.4 | 11 | 2347 | |||
XM_005254428.2 | XP_005254485.1 | 11 | 2219 | |||
XM_005254430.4 | XP_005254487.1 | 11 | 2141 | |||
XM_005254431.2 | XP_005254488.1 | 10 | 2102 | |||
XM_006720559.2 | XP_006720622.1 | 11 | 2222 | |||
XM_006720560.3 | XP_006720623.1 | 11 | 2318 | |||
XM_006720561.2 | XP_006720624.1 | 11 | 2136 | |||
XM_006720562.2 | XP_006720625.1 | 10 | 2105 | |||
XM_006720563.2 | XP_006720626.1 | 10 | 2084 | |||
XM_017022294.1 | XP_016877783.1 | 10 | 2024 | |||
XM_017022295.1 | XP_016877784.1 | 11 | 3014 | |||
GenBank mRNA | AF090915.1 | - | 11 | 2154 | ||
AF290612.1 | - | 11 | 2155 | |||
AK222819.1 | - | 11 | 2171 | |||
AL833611.1 | - | 11 | 2118 | |||
BC001308.1 | - | 11 | 2150 | 3453354 | ||
BC010838.1 | - | 9 | 1892 | |||
BC012887.1 | - | 11 | 2128 | 3882061 | ||
BC024772.1 | - | 11 | 2153 | 4342499 | ||
BG259258.1 | - | 267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
