siRNA Details
Gene Symbol: ACTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005386.2 | NP_001005386.1 | 10 | 1755 | ||
NM_005722.3 | NP_005713.1 | 9 | 1740 | |||
GenBank mRNA | AF006082.1 | - | 9 | 1599 | ||
BC014546.1 | - | 9 | 1606 | 4247021 | ||
BX649080.1 | - | 10 | 1613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
