siRNA Details
Gene Symbol: DNAJC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015268.3 | NP_056083.3 | 56 | 7167 | ||
XM_005247245.4 | XP_005247302.1 | 57 | 7364 | |||
XM_017006036.1 | XP_016861525.1 | 43 | 5624 | |||
GenBank mRNA | AB014578.2 | - | 56 | 7216 | ||
AK023168.1 | - | 25 | 3595 | |||
AK125330.1 | - | 33 | 4550 | |||
AK127112.1 | - | 40 | 5256 | |||
AY779857.1 | - | 56 | 7167 | |||
BC000164.3 | - | 9 | 1529 | |||
BC009630.1 | - | 11 | 1650 | |||
BC040638.2 | - | 6 | 1218 | |||
BC065756.1 | - | 5 | 927 | |||
BC105580.1 | - | 4 | 743 | |||
BC151246.1 | - | 57 | 7237 | 9007477 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
