siRNA Details
Gene Symbol: DNAJC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015268.3 | NP_056083.3 | 48, 49 | 5744 | ||
XM_005247245.4 | XP_005247302.1 | 49, 50 | 5941 | |||
XM_017006036.1 | XP_016861525.1 | 35, 36 | 4201 | |||
GenBank mRNA | AB014578.2 | - | 48, 49 | 5793 | ||
AK023168.1 | - | 17, 18 | 2172 | |||
AK125330.1 | - | 25, 26 | 3128 | |||
AK127112.1 | - | 32, 33 | 3832 | |||
AY779857.1 | - | 48, 49 | 5744 | |||
BC000164.3 | - | 1, 2 | 107 | |||
BC009630.1 | - | 3, 4 | 227 | |||
BC151246.1 | - | 49, 50 | 5814 | 9007477 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
