siRNA Details
Gene Symbol: DNAJC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015268.3 | NP_056083.3 | 52 | 6241 | ||
XM_005247245.4 | XP_005247302.1 | 53 | 6438 | |||
XM_017006036.1 | XP_016861525.1 | 39 | 4698 | |||
GenBank mRNA | AB014578.2 | - | 52 | 6290 | ||
AK023168.1 | - | 21 | 2669 | |||
AK125330.1 | - | 29 | 3625 | |||
AK127112.1 | - | 36 | 4329 | |||
AY779857.1 | - | 52 | 6241 | |||
BC000164.3 | - | 5 | 604 | |||
BC009630.1 | - | 7 | 724 | |||
BC040638.2 | - | 2 | 292 | |||
BC151246.1 | - | 53 | 6311 | 9007477 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
