siRNA Details
Gene Symbol: HECW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304840.1 | NP_001291769.1 | 27 | 6362 | ||
NM_020760.2 | NP_065811.1 | 29 | 6696 | |||
XM_006712646.3 | XP_006712709.1 | 29 | 6754 | |||
XM_006712648.3 | XP_006712711.1 | 28 | 6687 | |||
XM_017004563.1 | XP_016860052.1 | 29 | 6733 | |||
GenBank mRNA | AB037722.1 | - | 29 | 6696 | ||
AL390186.1 | - | 6 | 2461 | |||
CR749424.1 | - | 27 | 6460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
