siRNA Details
Gene Symbol: HEBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015987.4 | NP_057071.2 | 2 | 315 | ||
GenBank mRNA | AK289877.1 | - | 2 | 279 | ||
BC016277.2 | - | 2 | 150 | 3907035 | ||
BI550915.1 | - | 2 | 319 | |||
BT007294.1 | - | 2 | 120 | |||
CF126276.1 | - | 2 | 316 |
