siRNA Details
Gene Symbol: HECW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304840.1 | NP_001291769.1 | 22, 23 | 4003 | ||
NM_020760.2 | NP_065811.1 | 24, 25 | 4337 | |||
XM_006712646.3 | XP_006712709.1 | 24, 25 | 4395 | |||
XM_006712648.3 | XP_006712711.1 | 23, 24 | 4328 | |||
XM_017004563.1 | XP_016860052.1 | 24, 25 | 4374 | |||
GenBank mRNA | AB037722.1 | - | 24, 25 | 4337 | ||
AL390186.1 | - | 1, 2 | 102 | |||
BC117194.1 | - | 24, 25 | 4253 | 40125745 | ||
BC117198.1 | - | 24, 25 | 4253 | 40125749 | ||
CR749424.1 | - | 22, 23 | 4101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
