siRNA Details
Gene Symbol: LMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001395.2 | NP_001001395.1 | 4 | 688 | ||
NM_001243609.1 | NP_001230538.1 | 5 | 755 | |||
NM_001243610.1 | NP_001230539.1 | 5 | 527 | |||
NM_001243611.1 | NP_001230540.1 | 4 | 924 | |||
NM_001243612.1 | NP_001230541.1 | 4 | 612 | |||
NM_001243613.1 | NP_001230542.1 | 6 | 692 | |||
NM_018640.4 | NP_061110.2 | 4 | 946 | |||
NR_045012.1 | 6 | 880 | ||||
NR_045013.1 | 5 | 696 | ||||
NR_045014.1 | 4 | 629 | ||||
XM_006719110.3 | XP_006719173.1 | 5 | 763 | |||
XM_006719111.2 | XP_006719174.1 | 4 | 806 | |||
XM_011520762.1 | XP_011519064.1 | 5 | 654 | |||
XM_011520763.1 | XP_011519065.1 | 5 | 561 | |||
XM_017019643.1 | XP_016875132.1 | 5 | 1107 | |||
GenBank mRNA | AB044745.1 | - | 4 | 692 | ||
AB044746.1 | - | 6 | 593 | |||
AF258348.1 | - | 5 | 484 | |||
AK095595.1 | - | 4 | 564 | |||
AK294474.1 | - | 4 | 612 | |||
AK294909.1 | - | 6 | 692 | |||
AK308594.1 | - | 7 | 1040 | |||
AK308826.1 | - | 7 | 756 | |||
AK308896.1 | - | 6 | 692 | |||
AK312418.1 | - | 4 | 830 | |||
AK316021.1 | - | 4 | 548 | |||
BC017777.1 | - | 4 | 442 | |||
BC026311.1 | - | 4 | 842 | 4799836 | ||
BC050085.1 | - | 5 | 528 | 5274811 | ||
BI553855.1 | - | 4 | 585 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
