siRNA Details
Gene Symbol: LMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001395.2 | NP_001001395.1 | 4 | 3196 | ||
NM_001243609.1 | NP_001230538.1 | 5 | 3263 | |||
NM_001243610.1 | NP_001230539.1 | 5 | 3035 | |||
NM_001243611.1 | NP_001230540.1 | 4 | 3432 | |||
NM_001243612.1 | NP_001230541.1 | 4 | 3120 | |||
NM_001243613.1 | NP_001230542.1 | 6 | 3200 | |||
NM_018640.4 | NP_061110.2 | 4 | 3454 | |||
NR_045012.1 | 6 | 3388 | ||||
NR_045013.1 | 5 | 3204 | ||||
NR_045014.1 | 4 | 3137 | ||||
XM_006719110.3 | XP_006719173.1 | 5 | 3271 | |||
XM_006719111.2 | XP_006719174.1 | 4 | 3314 | |||
XM_011520762.1 | XP_011519064.1 | 5 | 3162 | |||
XM_011520763.1 | XP_011519065.1 | 5 | 3069 | |||
XM_017019643.1 | XP_016875132.1 | 5 | 3615 | |||
GenBank mRNA | AB044745.1 | - | 4 | 3200 | ||
AK095595.1 | - | 4 | 3070 | |||
AL050152.1 | - | 1 | 2491 | |||
AL050188.1 | - | 1 | 838 | |||
BC026311.1 | - | 4 | 3349 | 4799836 | ||
BC050085.1 | - | 5 | 3036 | 5274811 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
