siRNA Details
Gene Symbol: NAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004537.5 | NP_004528.1 | 14, 15 | 1621 | ||
NM_139207.3 | NP_631946.1 | 14, 15 | 1621 | |||
XM_006719425.3 | XP_006719488.1 | 12, 13 | 1084 | |||
XM_011538390.2 | XP_011536692.2 | 14, 15 | 1280 | |||
XM_011538393.2 | XP_011536695.1 | 14, 15 | 1533 | |||
XM_017019338.1 | XP_016874827.1 | 14, 15 | 1533 | |||
XM_017019339.1 | XP_016874828.1 | 12, 13 | 1344 | |||
XM_017019340.1 | XP_016874829.1 | 12, 13 | 1084 | |||
GenBank mRNA | AK122670.1 | - | 12, 13 | 1155 | ||
AK297500.1 | - | 11, 12 | 1093 | |||
AK298936.1 | - | 7, 8 | 562 | |||
AK304787.1 | - | 13, 14 | 1234 | |||
AL162068.1 | - | 14, 15 | 1258 | |||
BC002387.2 | - | 14, 15 | 1255 | 2964631 | ||
BC015599.1 | - | 13, 14 | 1470 | |||
BT007023.1 | - | 13, 14 | 1134 | |||
M86667.1 | - | 14, 15 | 1209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
