siRNA Details
Gene Symbol: MON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278469.1 | NP_001265398.1 | 26 | 3831 | ||
NM_001278470.1 | NP_001265399.1 | 26 | 3831 | |||
NM_001278471.1 | NP_001265400.1 | 26 | 3762 | |||
NM_001278472.1 | NP_001265401.1 | 27 | 3909 | |||
NM_015026.2 | NP_055841.2 | 26 | 3831 | |||
XM_017019041.1 | XP_016874530.1 | 27 | 3834 | |||
XM_017019042.1 | XP_016874531.1 | 27 | 3834 | |||
XM_017019043.1 | XP_016874532.1 | 28 | 3908 | |||
XM_017019044.1 | XP_016874533.1 | 28 | 3910 | |||
XM_017019045.1 | XP_016874534.1 | 29 | 3578 | |||
XM_017019046.1 | XP_016874535.1 | 23 | 3003 | |||
GenBank mRNA | AB017814.1 | - | 26 | 3556 | ||
AB028963.3 | - | 27 | 3802 | |||
AK022671.1 | - | 1 | 26 | |||
AK092646.1 | - | 1 | 613 | |||
AL833066.2 | - | 26 | 3826 | |||
AL834320.1 | - | 20 | 2789 | |||
BC136621.1 | - | 26 | 3800 | 9020611 | ||
BC144309.1 | - | 26 | 3731 | 9052829 | ||
BC151241.1 | - | 27 | 3825 | 8860514 | ||
BX537415.1 | - | 26 | 3829 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
