siRNA Details
Gene Symbol: MAGI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142782.1 | NP_001136254.1 | 16 | 2975 | ||
NM_152900.2 | NP_690864.2 | 16 | 2975 | |||
XM_005270737.3 | XP_005270794.1 | 16 | 2975 | |||
XM_011541208.2 | XP_011539510.1 | 16 | 4926 | |||
XM_017000974.1 | XP_016856463.1 | 16 | 3354 | |||
XM_017000975.1 | XP_016856464.1 | 10 | 1947 | |||
GenBank mRNA | AB046854.1 | - | 12 | 2041 | ||
AF213259.1 | - | 16 | 2794 | |||
AK026417.1 | - | 1 | 29 | |||
BC130409.1 | - | 16 | 2975 | 40146440 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
