siRNA Details
Gene Symbol: DNAJC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013238.2 | NP_037370.2 | 5 | 759 | ||
GenBank mRNA | AF126743.1 | - | 6 | 781 | ||
AJ270691.1 | - | 2 | 125 | |||
AK311866.1 | - | 5 | 395 | |||
AY248898.1 | - | 5 | 759 | |||
AY512565.1 | - | 5 | 452 | |||
BC010910.1 | - | 5 | 488 | 4332619 | ||
BC095400.1 | - | 5 | 378 | 30530999 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
