siRNA Details
Gene Symbol: DGKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204504.2 | NP_001191433.1 | 16 | 1756 | ||
NM_001204505.2 | NP_001191434.1 | 13 | 1377 | |||
NM_001204506.2 | NP_001191435.1 | 13 | 1377 | |||
NM_001297429.1 | NP_001284358.1 | 11 | 1144 | |||
NM_152910.5 | NP_690874.2 | 15 | 1719 | |||
NM_178009.4 | NP_821077.1 | 15 | 1719 | |||
NR_123714.1 | 13 | 1377 | ||||
NR_123715.1 | 15 | 1990 | ||||
GenBank mRNA | AB078967.1 | - | 15 | 1674 | ||
AB078968.1 | - | 15 | 1674 | |||
AK122909.1 | - | 13 | 1342 | |||
AK289546.1 | - | 16 | 1758 | |||
AK302631.1 | - | 11 | 1104 | |||
AK302727.1 | - | 13 | 1377 | |||
BC043292.2 | - | 13 | 1334 | 5297895 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
