siRNA Details
Gene Symbol: KIAA0556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015202.3 | NP_056017.3 | 13 | 1519 | ||
XM_005255201.3 | XP_005255258.1 | 14 | 1721 | |||
XM_005255202.3 | XP_005255259.1 | 14 | 1667 | |||
XM_005255203.3 | XP_005255260.1 | 13 | 1592 | |||
XM_005255206.3 | XP_005255263.1 | 14 | 1721 | |||
XM_005255208.2 | XP_005255265.1 | 4 | 419 | |||
XM_006721025.3 | XP_006721088.1 | 12 | 1460 | |||
XM_011545773.2 | XP_011544075.1 | 14 | 1691 | |||
XM_011545774.1 | XP_011544076.1 | 13 | 1637 | |||
XM_011545775.2 | XP_011544077.1 | 13 | 1601 | |||
XM_011545776.2 | XP_011544078.1 | 13 | 1589 | |||
XM_017023085.1 | XP_016878574.1 | 12 | 1472 | |||
XM_017023086.1 | XP_016878575.1 | 12 | 1472 | |||
XM_017023087.1 | XP_016878576.1 | 12 | 1406 | |||
XM_017023088.1 | XP_016878577.1 | 9 | 1193 | |||
GenBank mRNA | AB011128.2 | - | 13 | 1517 | ||
AK097485.1 | - | 10 | 1356 | |||
BC150276.1 | - | 13 | 1517 | 9007250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
