siRNA Details
Gene Symbol: CLEC16A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015226.2 | NP_056041.1 | 24 | 3917 | ||
XM_005255210.1 | XP_005255267.1 | 23 | 3905 | |||
XM_005255211.1 | XP_005255268.1 | 24 | 3863 | |||
XM_005255213.1 | XP_005255270.1 | 23 | 3857 | |||
XM_005255214.1 | XP_005255271.1 | 23 | 3746 | |||
XM_011522434.1 | XP_011520736.1 | 23 | 3782 | |||
XM_011522435.1 | XP_011520737.1 | 25 | 4035 | |||
XM_017023090.1 | XP_016878579.1 | 12 | 2196 | |||
GenBank mRNA | AB002348.3 | - | 24 | 3820 | ||
AK126771.1 | - | 4 | 1249 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
