siRNA Details
Gene Symbol: CLEC16A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015226.2 | NP_056041.1 | 11, 12 | 1540 | ||
XM_005255210.1 | XP_005255267.1 | 10, 11 | 1528 | |||
XM_005255214.1 | XP_005255271.1 | 11, 12 | 1534 | |||
XM_005255215.3 | XP_005255272.1 | 11, 12 | 1534 | |||
XM_005255216.1 | XP_005255273.1 | 11, 12 | 1534 | |||
XM_006720870.3 | XP_006720933.1 | 11, 12 | 1534 | |||
XM_011522434.1 | XP_011520736.1 | 11, 12 | 1534 | |||
XM_011522435.1 | XP_011520737.1 | 11, 12 | 1534 | |||
XM_011522436.2 | XP_011520738.1 | 11, 12 | 1534 | |||
XM_011522437.2 | XP_011520739.1 | 11, 12 | 1534 | |||
XM_011522438.2 | XP_011520740.1 | 11, 12 | 1534 | |||
XM_011522439.2 | XP_011520741.1 | 11, 12 | 1534 | |||
XM_011522440.2 | XP_011520742.1 | 11, 12 | 1534 | |||
GenBank mRNA | AB002348.3 | - | 11, 12 | 1443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
