siRNA Details
Gene Symbol: KDM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005366.1 | NP_001005366.1 | 14, 15 | 2424 | ||
NM_032590.4 | NP_115979.3 | 14, 15 | 2214 | |||
XM_005253955.3 | XP_005254012.1 | 14, 15 | 2424 | |||
XM_005253956.3 | XP_005254013.1 | 14, 15 | 2021 | |||
XM_005253961.4 | XP_005254018.1 | 3, 4 | 509 | |||
XM_011538867.2 | XP_011537169.1 | 14, 15 | 3467 | |||
XM_011538868.2 | XP_011537170.1 | 14, 15 | 3463 | |||
XM_011538869.2 | XP_011537171.1 | 14, 15 | 3465 | |||
XM_011538875.2 | XP_011537177.1 | 3, 4 | 509 | |||
GenBank mRNA | AB031230.1 | - | 4, 5 | 673 | ||
AJ459424.1 | - | 14, 15 | 2186 | |||
AK027440.1 | - | 4, 5 | 603 | |||
AK027692.1 | - | 1, 2 | 119 | |||
AK127328.1 | - | 14, 15 | 2424 | |||
AK299827.1 | - | 3, 4 | 519 | |||
AK316470.1 | - | 3, 4 | 521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
