siRNA Details
Gene Symbol: FAM214A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286495.1 | NP_001273424.1 | 12 | 4180 | ||
NM_019600.3 | NP_062546.2 | 13 | 4179 | |||
NR_104457.1 | 13 | 4416 | ||||
XM_005254547.2 | XP_005254604.1 | 13 | 4142 | |||
XM_005254548.2 | XP_005254605.1 | 13 | 4661 | |||
XM_011521789.2 | XP_011520091.1 | 13 | 4208 | |||
XM_011521790.1 | XP_011520092.1 | 11 | 4111 | |||
GenBank mRNA | AL832519.1 | - | 2 | 1517 | ||
BC040548.1 | - | 13 | 4416 | 5288206 | ||
BC058839.1 | - | 8 | 1975 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
