siRNA Details
Gene Symbol: SNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242933.1 | NP_001229862.1 | 9 | 995 | ||
NM_003099.4 | NP_003090.2 | 9 | 995 | |||
NM_148955.3 | NP_683758.1 | 7 | 800 | |||
XM_005254611.2 | XP_005254668.1 | 9 | 971 | |||
GenBank mRNA | AB209013.1 | - | 9 | 802 | ||
AF065483.1 | - | 9 | 865 | |||
AF065484.1 | - | 7 | 670 | |||
AK128179.1 | - | 9 | 887 | |||
AK222561.1 | - | 9 | 879 | |||
AK222793.1 | - | 9 | 868 | |||
AK291752.1 | - | 9 | 879 | |||
AK310618.1 | - | 6 | 679 | |||
BC000357.2 | - | 9 | 875 | 2964409 | ||
BQ935515.1 | - | 9 | 874 | |||
BT006983.1 | - | 9 | 865 | |||
U53225.1 | - | 9 | 910 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
