siRNA Details
Gene Symbol: SPAG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130527.2 | NP_001123999.1 | 20, 21 | 2790 | ||
NM_001130528.2 | NP_001124000.1 | 20, 21 | 2820 | |||
NM_001251971.1 | NP_001238900.1 | 16, 17 | 2448 | |||
NM_003971.5 | NP_003962.3 | 19, 20 | 2778 | |||
XM_005257768.2 | XP_005257825.1 | 20, 21 | 2820 | |||
XM_005257771.2 | XP_005257828.1 | 19, 20 | 2778 | |||
XM_005257774.3 | XP_005257831.1 | 16, 17 | 2364 | |||
XM_017025283.1 | XP_016880772.1 | 17, 18 | 2406 | |||
XM_017025284.1 | XP_016880773.1 | 17, 18 | 2406 | |||
XM_017025285.1 | XP_016880774.1 | 19, 20 | 2251 | |||
GenBank mRNA | AB011088.3 | - | 20, 21 | 2798 | ||
AF327452.1 | - | 20, 21 | 2659 | |||
AF542172.1 | - | 2, 3 | 261 | |||
AK024068.1 | - | 11, 12 | 1769 | |||
AK295098.1 | - | 16, 17 | 2358 | |||
AK299553.1 | - | 11, 12 | 1435 | |||
AK302789.1 | - | 20, 21 | 2699 | |||
AY219897.1 | - | 17, 18 | 2129 | |||
AY236487.1 | - | 6, 7 | 908 | |||
AY850123.1 | - | 19, 20 | 2753 | |||
BC146755.1 | - | 20, 21 | 2798 | 9007191 | ||
BC153878.1 | - | 20, 21 | 2809 | 9007194 | ||
X91879.1 | - | 18, 19 | 2184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
