siRNA Details
Gene Symbol: MMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012329.2 | NP_036461.2 | 7 | 988 | ||
XM_006721795.2 | XP_006721858.1 | 8 | 1094 | |||
GenBank mRNA | AK303810.1 | - | 7 | 833 | ||
AK312760.1 | - | 7 | 867 | |||
AY424289.1 | - | 7 | 968 | |||
BC026324.1 | - | 7 | 960 | 4794375 | ||
CR541893.1 | - | 7 | 703 | |||
X85750.1 | - | 7 | 784 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
