siRNA Details
Gene Symbol: ANXA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003954.2 | NP_001003954.1 | 6, 7 | 617 | ||
NM_004306.3 | NP_004297.2 | 5, 6 | 494 | |||
GenBank mRNA | AJ306450.1 | - | 6, 7 | 593 | ||
AK223374.1 | - | 5, 6 | 493 | |||
AK290281.1 | - | 6, 7 | 603 | |||
BC125158.1 | - | 5, 6 | 464 | 40120955 | ||
CR541838.1 | - | 4, 5 | 402 | |||
Z11502.1 | - | 5, 6 | 481 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
