siRNA Details
Gene Symbol: ANXA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003954.2 | NP_001003954.1 | 4 | 341 | ||
NM_004306.3 | NP_004297.2 | 3 | 218 | |||
GenBank mRNA | AJ306450.1 | - | 4 | 317 | ||
AK223374.1 | - | 3 | 217 | |||
AK290281.1 | - | 4 | 327 | |||
BC125158.1 | - | 3 | 188 | 40120955 | ||
CR541838.1 | - | 2 | 126 | |||
Z11502.1 | - | 3 | 205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
