siRNA Details
Gene Symbol: TCEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007108.3 | NP_009039.1 | 2, 3 | 200 | ||
NM_207013.2 | NP_996896.1 | 2, 3 | 200 | |||
GenBank mRNA | BC013306.2 | - | 1, 2 | 154 | 2963043 | |
BC065000.1 | - | 2, 3 | 156 | 6018611 | ||
BF967495.2 | - | 2, 3 | 206 | |||
BG329435.1 | - | 181 | ||||
BM553320.1 | - | 2, 3 | 215 | |||
BM700019.1 | - | 1, 2 | 151 | |||
BM921799.1 | - | 1, 2 | 168 | |||
BU533433.1 | - | 2, 3 | 171 | |||
DT217333.1 | - | 2, 3 | 176 | |||
L42856.1 | - | 1, 2 | 143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
