siRNA Details
Gene Symbol: WSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015626.8 | NP_056441.6 | 9 | 2316 | ||
NM_134265.2 | NP_599027.1 | 7 | 1878 | |||
XM_005257963.3 | XP_005258020.1 | 8 | 2113 | |||
XM_017024433.1 | XP_016879922.1 | 8 | 2202 | |||
XM_017024434.1 | XP_016879923.1 | 10 | 2404 | |||
XM_017024435.1 | XP_016879924.1 | 10 | 3771 | |||
XM_017024436.1 | XP_016879925.1 | 8 | 3503 | |||
GenBank mRNA | AK074917.1 | - | 9 | 2282 | ||
AK091514.1 | - | 2481 | ||||
AL110269.1 | - | 9 | 3642 | |||
BC005226.1 | - | 4 | 1321 | |||
BC048007.1 | - | 9 | 3638 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
