siRNA Details
Gene Symbol: WSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015626.8 | NP_056441.6 | 9 | 1916 | ||
NM_134265.2 | NP_599027.1 | 7 | 1478 | |||
XM_005257963.3 | XP_005258020.1 | 8 | 1713 | |||
XM_017024433.1 | XP_016879922.1 | 8 | 1802 | |||
XM_017024434.1 | XP_016879923.1 | 10 | 2004 | |||
XM_017024435.1 | XP_016879924.1 | 10 | 3371 | |||
XM_017024436.1 | XP_016879925.1 | 8 | 3103 | |||
GenBank mRNA | AF069313.2 | - | 9 | 1865 | ||
AF072880.1 | - | 9 | 1690 | |||
AF106683.1 | - | 9 | 1733 | |||
AF106684.1 | - | 4 | 1465 | |||
AF112205.1 | - | 9 | 1855 | |||
AF240696.1 | - | 7 | 1427 | |||
AK074917.1 | - | 9 | 1882 | |||
AK222576.1 | - | 9 | 1882 | |||
AL110243.1 | - | 9 | 1916 | |||
AL110269.1 | - | 9 | 3243 | |||
BC005226.1 | - | 4 | 918 | |||
BC021110.1 | - | 9 | 1863 | 4865448 | ||
BC048007.1 | - | 9 | 3238 | |||
BU189299.1 | - | 1 | 149 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
