siRNA Details
Gene Symbol: WSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015626.8 | NP_056441.6 | 9 | 1639 | ||
NM_134265.2 | NP_599027.1 | 7 | 1201 | |||
XM_005257963.3 | XP_005258020.1 | 8 | 1436 | |||
XM_017024433.1 | XP_016879922.1 | 8 | 1525 | |||
XM_017024434.1 | XP_016879923.1 | 10 | 1727 | |||
XM_017024435.1 | XP_016879924.1 | 10 | 3094 | |||
XM_017024436.1 | XP_016879925.1 | 8 | 2826 | |||
GenBank mRNA | AF069313.2 | - | 9 | 1588 | ||
AF072880.1 | - | 9 | 1414 | |||
AF106683.1 | - | 9 | 1456 | |||
AF106684.1 | - | 4 | 1188 | |||
AF112205.1 | - | 9 | 1578 | |||
AF240696.1 | - | 7 | 1150 | |||
AK074917.1 | - | 9 | 1605 | |||
AK222576.1 | - | 9 | 1605 | |||
AL110243.1 | - | 9 | 1639 | |||
AL110269.1 | - | 9 | 2966 | |||
BC005226.1 | - | 4 | 641 | |||
BC021110.1 | - | 9 | 1586 | 4865448 | ||
BC048007.1 | - | 9 | 2961 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
