siRNA Details
Gene Symbol: SETD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014712.2 | NP_055527.1 | 4 | 1026 | ||
XM_005255723.1 | XP_005255780.1 | 4 | 510 | |||
XM_006721106.3 | XP_006721169.1 | 4 | 431 | |||
XM_017023909.1 | XP_016879398.1 | 4 | 1663 | |||
GenBank mRNA | AB002337.2 | - | 4 | 1026 | ||
BC035795.2 | - | 4 | 526 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
