siRNA Details
Gene Symbol: OGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014057.4 | NP_054776.1 | 4 | 662 | ||
NM_024416.4 | NP_077727.3 | 4 | 581 | |||
NM_033014.3 | NP_148935.1 | 4 | 944 | |||
GenBank mRNA | AF086912.1 | - | 4 | 509 | ||
AF100758.1 | - | 4 | 509 | |||
AF202167.1 | - | 3 | 434 | |||
AK075521.1 | - | 3 | 473 | |||
AK129845.1 | - | 3 | 466 | |||
AK289628.1 | - | 4 | 659 | |||
AK295433.1 | - | 4 | 581 | |||
AL110267.1 | - | 3 | 451 | |||
BC037273.2 | - | 3 | 450 | 5265880 | ||
BC095443.1 | - | 3 | 466 | 30405707 | ||
BG772245.1 | - | 4 | 585 | |||
BX646984.1 | - | 4 | 619 | |||
CR533516.1 | - | 3 | 383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
