siRNA Details
Gene Symbol: OGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014057.4 | NP_054776.1 | 4, 5 | 708 | ||
NM_024416.4 | NP_077727.3 | 4, 5 | 627 | |||
NM_033014.3 | NP_148935.1 | 4, 5 | 990 | |||
GenBank mRNA | AF086912.1 | - | 4, 5 | 555 | ||
AF088023.1 | - | 1, 2 | 46 | |||
AF100758.1 | - | 4, 5 | 555 | |||
AF202167.1 | - | 3, 4 | 480 | |||
AK075521.1 | - | 3, 4 | 519 | |||
AK129845.1 | - | 3, 4 | 512 | |||
AK289628.1 | - | 4, 5 | 705 | |||
AK295433.1 | - | 4, 5 | 627 | |||
AL110267.1 | - | 3, 4 | 497 | |||
BC037273.2 | - | 3, 4 | 496 | 5265880 | ||
BC095443.1 | - | 3, 4 | 512 | 30405707 | ||
BG772245.1 | - | 4, 5 | 631 | |||
BX646984.1 | - | 4, 5 | 665 | |||
CR533516.1 | - | 3, 4 | 429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
