siRNA Details
Gene Symbol: OGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014057.4 | NP_054776.1 | 6 | 979 | ||
NM_024416.4 | NP_077727.3 | 6 | 898 | |||
NM_033014.3 | NP_148935.1 | 6 | 1261 | |||
GenBank mRNA | AF086912.1 | - | 6 | 826 | ||
AF088023.1 | - | 3 | 317 | |||
AF100758.1 | - | 6 | 826 | |||
AF202167.1 | - | 5 | 751 | |||
AK075521.1 | - | 5 | 790 | |||
AK129845.1 | - | 5 | 783 | |||
AK289628.1 | - | 6 | 976 | |||
AK295433.1 | - | 6 | 898 | |||
AL110267.1 | - | 5 | 768 | |||
AL133114.1 | - | 2 | 244 | |||
BC037273.2 | - | 5 | 767 | 5265880 | ||
BC095443.1 | - | 5 | 783 | 30405707 | ||
CR533516.1 | - | 5 | 700 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
