siRNA Details
Gene Symbol: DOK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018431.4 | NP_060901.2 | 5 | 926 | ||
NM_177959.2 | NP_808874.1 | 5 | 572 | |||
XM_011528904.1 | XP_011527206.1 | 5 | 555 | |||
GenBank mRNA | AF132732.1 | - | 5 | 648 | ||
AF466368.1 | - | 5 | 451 | |||
AK313222.1 | - | 5 | 638 | |||
AL050069.1 | - | 4 | 329 | |||
BC008992.1 | - | 5 | 572 | 4183000 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
