siRNA Details
Gene Symbol: TYROBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173514.1 | NP_001166985.1 | 4 | 356 | ||
NM_001173515.1 | NP_001166986.1 | 4 | 353 | |||
NM_003332.3 | NP_003323.1 | 5 | 389 | |||
NM_198125.2 | NP_937758.1 | 5 | 386 | |||
NR_033390.1 | 4 | 349 | ||||
GenBank mRNA | AF019562.1 | - | 5 | 375 | ||
AJ010098.1 | - | 5 | 307 | |||
AK290385.1 | - | 5 | 372 | |||
AY074782.1 | - | 5 | 306 | |||
BC011175.1 | - | 5 | 364 | 4150408 | ||
BI908458.1 | - | 4 | 320 | |||
BP295666.1 | - | 4 | 322 | |||
BP302545.1 | - | 5 | 389 | |||
BP305672.1 | - | 4 | 340 | |||
BT009851.1 | - | 5 | 308 | |||
CR450342.1 | - | 5 | 308 | |||
CR542202.1 | - | 5 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
