siRNA Details
Gene Symbol: TYROBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173514.1 | NP_001166985.1 | 4 | 405 | ||
NM_001173515.1 | NP_001166986.1 | 4 | 402 | |||
NM_003332.3 | NP_003323.1 | 5 | 438 | |||
NM_198125.2 | NP_937758.1 | 5 | 435 | |||
NR_033390.1 | 4 | 398 | ||||
GenBank mRNA | AF019562.1 | - | 5 | 424 | ||
AK290385.1 | - | 5 | 421 | |||
BC011175.1 | - | 5 | 413 | 4150408 | ||
BI908458.1 | - | 4 | 369 | |||
BP295666.1 | - | 4 | 371 | |||
BP302545.1 | - | 5 | 438 | |||
BP305672.1 | - | 4 | 389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
