siRNA Details
Gene Symbol: HECTD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024602.5 | NP_078878.3 | 21 | 3350 | ||
XM_011542140.2 | XP_011540442.1 | 19 | 2747 | |||
GenBank mRNA | AK056545.1 | - | 19 | 2701 | ||
AK056873.1 | - | 13 | 2244 | |||
AK091583.1 | - | 6 | 2099 | |||
AK096462.1 | - | 21 | 3229 | |||
BC019105.2 | - | 6 | 1309 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
