siRNA Details
Gene Symbol: CYTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292018.1 | NP_001278947.1 | 14 | 3496 | ||
NM_001292019.1 | NP_001278948.1 | 14 | 3062 | |||
NM_004762.3 | NP_004753.1 | 14 | 3099 | |||
NM_017456.3 | NP_059430.2 | 13 | 3096 | |||
XM_011525475.2 | XP_011523777.1 | 13 | 3839 | |||
XM_011525476.2 | XP_011523778.1 | 14 | 3014 | |||
XM_011525477.1 | XP_011523779.1 | 12 | 2877 | |||
XM_011525478.1 | XP_011523780.1 | 13 | 3418 | |||
XM_017025336.1 | XP_016880825.1 | 13 | 3166 | |||
XM_017025338.1 | XP_016880827.1 | 13 | 3482 | |||
GenBank mRNA | AF054997.1 | - | 1 | 1459 | ||
AK091369.1 | - | 1 | 2598 | |||
AK123894.1 | - | 1 | 4162 | |||
AL832791.1 | - | 1 | 3275 | |||
BC038385.1 | - | 14 | 3019 | 4755203 | ||
BC050452.1 | - | 14 | 3010 | 6043039 | ||
M85169.1 | - | 14 | 3043 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
