siRNA Details
Gene Symbol: MYH7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020884.4 | NP_065935.3 | 8 | 973 | ||
XM_006723840.3 | XP_006723903.1 | 6 | 614 | |||
XM_011528941.2 | XP_011527243.1 | 6 | 614 | |||
XM_017027986.1 | XP_016883475.1 | 6 | 614 | |||
XM_017027987.1 | XP_016883476.1 | 2 | 226 | |||
GenBank mRNA | AB040945.2 | - | 6 | 682 | ||
AK000947.1 | - | 4 | 550 | |||
BC069327.1 | - | 4 | 533 | |||
BC069406.1 | - | 4 | 533 | |||
BC151242.1 | - | 6 | 694 | 8860515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
