siRNA Details
Gene Symbol: TBC1D9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015130.2 | NP_055945.2 | 21 | 3544 | ||
GenBank mRNA | AB020689.2 | - | 21 | 3347 | ||
AB449892.1 | - | 21 | 3272 | |||
BC038804.1 | - | 12 | 1658 | |||
BC146758.1 | - | 21 | 3347 | 9007196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
