siRNA Details
Gene Symbol: OPTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008211.1 | NP_001008212.1 | 4 | 548 | ||
NM_001008212.1 | NP_001008213.1 | 3 | 479 | |||
NM_001008213.1 | NP_001008214.1 | 4 | 533 | |||
NM_021980.4 | NP_068815.2 | 2 | 327 | |||
GenBank mRNA | AF061034.1 | - | 4 | 421 | ||
AF420371.1 | - | 4 | 405 | |||
AF420372.1 | - | 2 | 184 | |||
AF420373.1 | - | 3 | 336 | |||
AK055403.1 | - | 3 | 768 | |||
AK303715.1 | - | 2 | 251 | |||
AK316171.1 | - | 2 | 273 | |||
BC013876.2 | - | 2 | 240 | 3831267 | ||
BC032762.1 | - | 4 | 453 | 3457195 | ||
KU178502.1 | - | 1 | 95 | |||
KU178503.1 | - | 1 | 95 | |||
KU178504.1 | - | 1 | 95 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
